Methyl benzo[d]thiazole-7-carboxylate
Lieferant: Apollo Scientific
OR305041-250MG
APOSOR305041-250MGEA
0
EUR
APOSOR305041-250MG
Methyl benzo[d]thiazole-7-carboxylate
Methyl benzo[d]thiazole-7-carboxylate
Formel:
C₉H₇NO₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD13190218 CAS-Nummer: 1038509-28-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...