Benzo[d]thiazole-7-carboxylic acid
Lieferant: Apollo Scientific
OR305043-250MG
APOSOR305043-250MGEA
0
EUR
APOSOR305043-250MG
Benzo[d]thiazole-7-carboxylic acid
Benzo[d]thiazole-7-carboxylic acid
Formel:
C₈H₅NO₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD13195419 CAS-Nummer: 677304-83-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...