5-Methoxy-1-benzothiophene-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Methoxybenzo[b]thiophene-2-carboxylic acid

OR305046-250MG OR305046-1G
APOSOR305046-250MGEA 119 EUR
APOSOR305046-250MG APOSOR305046-1G
5-Methoxy-1-benzothiophene-2-carboxylic acid
5-Methoxy-1-benzothiophene-2-carboxylic acid
Formel: C₁₀H₈O₃S
Molecular Weight: 208.234 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07371536
CAS-Nummer: 23046-02-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR