6-Methoxy-1-benzothiophene-2-carboxylic acid

Lieferant: Apollo Scientific

Synonyme: 6-Methoxybenzo[b]thiophene-2-carboxylic acid

OR305047-250MG OR305047-1G
APOSOR305047-250MGEA 72.5 EUR
APOSOR305047-250MG APOSOR305047-1G
6-Methoxy-1-benzothiophene-2-carboxylic acid
6-Methoxy-1-benzothiophene-2-carboxylic acid
Formel: C₁₀H₈O₃S
Molecular Weight: 208.234 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07371542
CAS-Nummer: 102539-79-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR