1-Benzothiophen-6-ylmethanol

Lieferant: Apollo Scientific
Achtung

Synonyme: Benzo[b]thiophen-6-ylmethanol

OR305048-250MG
APOSOR305048-250MGEA 114 EUR
APOSOR305048-250MG
1-Benzothiophen-6-ylmethanol
1-Benzothiophen-6-ylmethanol
Formel: C₉H₈OS
Molecular Weight: 164,224 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18451826
CAS-Nummer: 6179-28-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR