2,6-Dichloro-4-methyl-3-nitropyridine 98%

Lieferant: Apollo Scientific
Achtung

OR305051-1G
APOSOR305051-1GEA 42.2 EUR
APOSOR305051-1G
2,6-Dichloro-4-methyl-3-nitropyridine 98%
2,6-Dichloro-4-methyl-3-nitropyridine
Formel: C₆H₄Cl₂N₂O₂
Molecular Weight: 207,01 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11110694
CAS-Nummer: 60010-03-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR