tert-Butyl 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole-1-carboxylate
Lieferant: Apollo Scientific
OR305068-250MG
APOSOR305068-250MGEA
66.7
EUR
APOSOR305068-250MG
tert-Butyl 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole-1-carboxylate
tert-Butyl 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole-1-carboxylate
Formel:
C₁₈H₂₅BN₂O₄ Molecular Weight: 344,213 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13183032 CAS-Nummer: 890839-29-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...