5,6-Dichlor-1H-indazol

Lieferant: Apollo Scientific
Achtung

OR305085-250MG
APOSOR305085-250MGEA 33.5 EUR
APOSOR305085-250MG
5,6-Dichlor-1H-indazol
5,6-Dichloro-1H-indazole
Formel: C₇H₄Cl₂N₂
Molecular Weight: 187,0300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11878111
CAS-Nummer: 124691-76-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR