5-Methyl-1H-indazol
Lieferant: Apollo Scientific
|
Achtung
|
OR305086-1G
OR305086-5G
OR305086-10G
APOSOR305086-1GEA
24.8
EUR
APOSOR305086-1G
APOSOR305086-5G
APOSOR305086-10G
5-Methyl-1H-indazol
5-Methyl-1H-indazole
Formel:
C₈H₈N₂ Molecular Weight: 132.1600 g/mol Siedepunkt: 165…190 °C/1.5mmHg Schmelzpunkt: 111 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01606235 CAS-Nummer: 1776-37-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...