4-Methoxy-1H-indazol

Lieferant: Apollo Scientific
Achtung

OR305095-1G
APOSOR305095-1GEA 130 EUR
APOSOR305095-1G
4-Methoxy-1H-indazol
4-Methoxy-1H-indazole
Formel: C₈H₈N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD09261131
CAS-Nummer: 351210-06-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR