tert-Butyl 4-bromo-1H-indazole-5-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-5-(tert-butoxycarbonyl)-1H-indazole

OR305101-100MG
APOSOR305101-100MGEA 0 EUR
APOSOR305101-100MG
tert-Butyl 4-bromo-1H-indazole-5-carboxylate
tert-Butyl 4-bromo-1H-indazole-5-carboxylate
Formel: C₁₂H₁₃BrN₂O₂
Molecular Weight: 297,148 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22572308
CAS-Nummer: 1203662-37-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR