4-Bromo-6-methoxy-1H-indazole
Lieferant: Apollo Scientific
OR305103-250MG
APOSOR305103-250MGEA
54.1
EUR
APOSOR305103-250MG
4-Bromo-6-methoxy-1H-indazole
4-Bromo-6-methoxy-1H-indazole
Formel:
C₈H₇BrN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD08272208 CAS-Nummer: 885520-83-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...