Methyl 5-bromo-1H-indazole-4-carboxylate

Lieferant: Apollo Scientific

OR305114-250MG
APOSOR305114-250MGEA 75.4 EUR
APOSOR305114-250MG
Methyl 5-bromo-1H-indazole-4-carboxylate
Methyl 5-bromo-1H-indazole-4-carboxylate
Formel: C₉H₇BrN₂O₂
Molecular Weight: 255,068 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13177017
CAS-Nummer: 1037840-79-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR