5-Bromo-7-(propan-2-yl)-1H-indazole

Lieferant: Apollo Scientific

Synonyme: 5-Bromo-7-isopropyl-1H-indazole

OR305118-250MG
APOSOR305118-250MGEA 36.4 EUR
APOSOR305118-250MG
5-Bromo-7-(propan-2-yl)-1H-indazole
5-Bromo-7-(propan-2-yl)-1H-indazole
Formel: C₁₀H₁₁BrN₂
Molecular Weight: 239,112 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13177024
CAS-Nummer: 773887-09-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR