5-Bromo-7-ethyl-1H-indazole
Lieferant: Apollo Scientific
|
Achtung
|
OR305119-250MG
APOSOR305119-250MGEA
56.7
EUR
APOSOR305119-250MG
5-Bromo-7-ethyl-1H-indazole
5-Bromo-7-ethyl-1H-indazole
Formel:
C₉H₉BrN₂ Molecular Weight: 225,085 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11044594 CAS-Nummer: 635712-49-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...