tert-Butyl 6-bromo-4-methoxy-1H-indazole-1-carboxylate

Lieferant: Apollo Scientific

OR305133-250MG
APOSOR305133-250MGEA 215 EUR
APOSOR305133-250MG
tert-Butyl 6-bromo-4-methoxy-1H-indazole-1-carboxylate
tert-Butyl 6-bromo-4-methoxy-1H-indazole-1-carboxylate
Formel: C₁₃H₁₅BrN₂O₃
Molecular Weight: 327,174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23134666
CAS-Nummer: 1169789-29-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR