6-Bromo-4-methoxy-1H-indazole

Lieferant: Apollo Scientific
Achtung

OR305141-1G
APOSOR305141-1GEA 58 EUR
APOSOR305141-1G
6-Bromo-4-methoxy-1H-indazole
6-Bromo-4-methoxy-1H-indazole
Formel: C₈H₇BrN₂O
Molecular Weight: 227,058 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07781361
CAS-Nummer: 885519-21-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR