Imidazo[1,2-a]pyridine-3-sulphonate

Lieferant: Apollo Scientific

OR305156-250MG OR305156-1G
APOSOR305156-250MGEA 223 EUR
APOSOR305156-250MG APOSOR305156-1G
Imidazo[1,2-a]pyridine-3-sulphonate
Imidazo[1,2-a]pyridine-3-sulphonate
Formel: C₇H₆N₂O₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD13176742
CAS-Nummer: 112581-51-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR