3-Iodimidazo[1,2-a]pyridin 95+%

Lieferant: Apollo Scientific

OR305157-5G
APOSOR305157-5GEA 121 EUR
APOSOR305157-5G
3-Iodimidazo[1,2-a]pyridin 95+%
3-Iodimidazo[1,2-a]pyridin
Formel: C₇H₅IN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06739222
CAS-Nummer: 307503-19-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR