Imidazo[1,2-a]pyridin-5-ylmethanol 95+%

Lieferant: Apollo Scientific

OR305161-1G
APOSOR305161-1GEA 156 EUR
APOSOR305161-1G
Imidazo[1,2-a]pyridin-5-ylmethanol 95+%
Imidazo[1,2-a]pyridin-5-ylmethanol
Formel: C₈H₈N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD13183181
CAS-Nummer: 167884-17-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR