6-Aminoimidazo[1,2-a]pyridin 95%

Lieferant: Apollo Scientific
Achtung

OR305164-100MG OR305164-1G OR305164-5G
APOSOR305164-100MGEA 21.9 EUR
APOSOR305164-100MG APOSOR305164-1G APOSOR305164-5G
6-Aminoimidazo[1,2-a]pyridin 95%
6-Aminoimidazo[1,2-a]pyridin
Formel: C₇H₇N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD06739232
CAS-Nummer: 235106-53-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR