7-Iodoimidazo[1,2-a]pyridine 95%

Lieferant: Apollo Scientific

OR305166-1G OR305166-250MG
APOSOR305166-1GEA 229 EUR
APOSOR305166-1G APOSOR305166-250MG
7-Iodoimidazo[1,2-a]pyridine 95%
7-Iodoimidazo[1,2-a]pyridine
Formel: C₇H₅IN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD13177252
CAS-Nummer: 908269-30-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR