2,3-Dihydro-1H-indole-7-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Carboxy-2,3-dihydro-1H-indole

OR305167-250MG
APOSOR305167-250MGEA 143 EUR
APOSOR305167-250MG
2,3-Dihydro-1H-indole-7-carboxylic acid
2,3-Dihydro-1H-indole-7-carboxylic acid
Formel: C₉H₉NO₂
Molecular Weight: 163,173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11617212
CAS-Nummer: 15861-40-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR