2-(2-Bromophenyl)-1H-imidazole

Lieferant: Apollo Scientific
Achtung

OR305175-5G
APOSOR305175-5GEA 177 EUR
APOSOR305175-5G
2-(2-Bromophenyl)-1H-imidazole
2-(2-Bromophenyl)-1H-imidazole
Formel: C₉H₇BrN₂
Molecular Weight: 223.069 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08668745
CAS-Nummer: 162356-38-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR