Methyl 1H-imidazo[4,5-b]pyridine-6-carboxylate

Lieferant: Apollo Scientific

OR305190-250MG
APOSOR305190-250MGEA 36.4 EUR
APOSOR305190-250MG
Methyl 1H-imidazo[4,5-b]pyridine-6-carboxylate
Methyl 1H-imidazo[4,5-b]pyridine-6-carboxylate
Formel: C₈H₇N₃O₂
Molecular Weight: 177,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13176754
CAS-Nummer: 77862-95-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR