2-Iodo-1-methyl-1H-indole-3-carbonitrile
Lieferant: Apollo Scientific
OR305196-250MG
APOSOR305196-250MGEA
734
EUR
APOSOR305196-250MG
2-Iodo-1-methyl-1H-indole-3-carbonitrile
2-Iodo-1-methyl-1H-indole-3-carbonitrile
Formel:
C₁₀H₇IN₂ Molecular Weight: 282,08 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13177409 CAS-Nummer: 490039-77-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...