2-Iodo-1H-indole

Lieferant: Apollo Scientific
Achtung

OR305199-1G
APOSOR305199-1GEA 329 EUR
APOSOR305199-1G
2-Iodo-1H-indole
2-Iodo-1H-indole
Formel: C₈H₆IN
Molecular Weight: 243.044 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD10699391
CAS-Nummer: 26340-49-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR