1H-Indole-5-carbohydrazide

Lieferant: Apollo Scientific

OR305214-250MG
APOSOR305214-250MGEA 174 EUR
APOSOR305214-250MG
1H-Indole-5-carbohydrazide
1H-Indole-5-carbohydrazide
Formel: C₉H₉N₃O
Molecular Weight: 175,187 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11977800
CAS-Nummer: 406192-82-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR