1H-Indole-5-carbonyl chloride

Lieferant: Apollo Scientific
Gefahr

OR305216-250MG
APOSOR305216-250MGEA 184 EUR
APOSOR305216-250MG
1H-Indole-5-carbonyl chloride
1H-Indole-5-carbonyl chloride
Formel: C₉H₆ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD11520769
CAS-Nummer: 161397-68-8
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR