1H-Indol-5-ylacetonitrile

Lieferant: Apollo Scientific

Synonyme: 2-(1H-Indol-5-yl)acetonitrile

OR305220-250MG
APOSOR305220-250MGEA 81.2 EUR
APOSOR305220-250MG
1H-Indol-5-ylacetonitrile
1H-Indol-5-ylacetonitrile
Formel: C₁₀H₈N₂
Molecular Weight: 156.184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09263862
CAS-Nummer: 23690-49-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR