1H-Indole-6-carbohydrazide

Lieferant: Apollo Scientific

OR305234-250MG
APOSOR305234-250MGEA 92.8 EUR
APOSOR305234-250MG
1H-Indole-6-carbohydrazide
1H-Indole-6-carbohydrazide
Formel: C₉H₉N₃O
Molecular Weight: 175,187 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11977799
CAS-Nummer: 851211-74-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR