1H-Indole-7-carboxamide 95%

Lieferant: Apollo Scientific

OR305242-1G
APOSOR305242-1GEA 46.4 EUR
APOSOR305242-1G
1H-Indole-7-carboxamide 95%
1H-Indole-7-carboxamide
Formel: C₉H₈N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00572866
CAS-Nummer: 1670-89-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR