1-tert-Butyl 6-methyl 2,3-dihydro-1H-indole-1,6-dicarboxylate

Lieferant: Apollo Scientific

Synonyme: 1-tert-Butyl 6-methyl indoline-1,6-dicarboxylate

OR305249-250MG
APOSOR305249-250MGEA 43.5 EUR
APOSOR305249-250MG
1-tert-Butyl 6-methyl 2,3-dihydro-1H-indole-1,6-dicarboxylate
1-tert-Butyl 6-methyl 2,3-dihydro-1H-indole-1,6-dicarboxylate
Formel: C₁₅H₁₉NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD09878912
CAS-Nummer: 1220039-51-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR