6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)indoline
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Indoline-6-boronic acid pinacol ester, 2,3-Dihydro-1H-indol-6-boronic acid pinacol ester, 6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)indoline, 2,3-Dihydro-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
OR305264-250MG
APOSOR305264-250MGEA
323
EUR
APOSOR305264-250MG
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)indoline
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)indoline
Formel:
C₁₄H₂₀BNO₂ Molecular Weight: 245,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16995802 CAS-Nummer: 2304634-69-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...