4-Hydroxyisoindolin-1-one
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,3-Dihydro-4-hydroxy-1H-isoindol-1-one, 4-Hydroxy-2,3-dihydroisoindol-1-one, 2,3-Dihydro-4-hydroxy-1-oxo-1H-isoindole
OR305268-250MG
APOSOR305268-250MGEA
31.9
EUR
APOSOR305268-250MG
4-Hydroxyisoindolin-1-one
4-Hydroxyisoindolin-1-one
Formel:
C₈H₇NO₂ Molecular Weight: 149,147 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10000829 CAS-Nummer: 366453-21-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...