6-Hydroxyisoindolin-1-one
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,3-Dihydro-6-hydroxy-1-oxo-1H-isoindole, 2,3-Dihydro-6-hydroxy-1H-isoindol-1-one
OR305269-250MG
APOSOR305269-250MGEA
50.9
EUR
APOSOR305269-250MG
6-Hydroxyisoindolin-1-one
6-Hydroxyisoindolin-1-one
Formel:
C₈H₇NO₂ Molecular Weight: 149,147 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08276013 CAS-Nummer: 659737-57-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...