4,6-Dichloro-2,3-dihydro-1H-isoindol-1-one 95%

Lieferant: Apollo Scientific

Synonyme: 4,6-Dichloroisoindolin-1-one

OR305276-250MG
APOSOR305276-250MGEA 112 EUR
APOSOR305276-250MG
4,6-Dichloro-2,3-dihydro-1H-isoindol-1-one 95%
4,6-Dichloro-2,3-dihydro-1H-isoindol-1-one
Formel: C₈H₅Cl₂NO
Molecular Weight: 202,037 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13177812
CAS-Nummer: 74572-30-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR