5-(Hydroxymethyl)-2,3-dihydro-1H-isoindol-1-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(Hydroxymethyl)isoindolin-1-one

OR305277-250MG
APOSOR305277-250MGEA 389 EUR
APOSOR305277-250MG
5-(Hydroxymethyl)-2,3-dihydro-1H-isoindol-1-one
5-(Hydroxymethyl)-2,3-dihydro-1H-isoindol-1-one
Formel: C₉H₉NO₂
Molecular Weight: 163,173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13177814
CAS-Nummer: 926307-97-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR