6-Iodo-2,3-dihydro-1H-isoindol-1-one

Lieferant: Apollo Scientific

Synonyme: 6-Iodoisoindolin-1-one

OR305289-250MG
APOSOR305289-250MGEA 101 EUR
APOSOR305289-250MG
6-Iodo-2,3-dihydro-1H-isoindol-1-one
6-Iodo-2,3-dihydro-1H-isoindol-1-one
Formel: C₈H₆INO
Molecular Weight: 259,044 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13177830
CAS-Nummer: 675109-30-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR