6-Nitro-2,3-dihydro-1H-isoindol-1-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Nitroisoindolin-1-one

OR305291-250MG
APOSOR305291-250MGEA 21.9 EUR
APOSOR305291-250MG
6-Nitro-2,3-dihydro-1H-isoindol-1-one
6-Nitro-2,3-dihydro-1H-isoindol-1-one
Formel: C₈H₆N₂O₃
Molecular Weight: 178,145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03093931
CAS-Nummer: 110568-64-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR