6-Chloro-2,3-dihydro-1H-isoindol-1-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Chloroisoindolin-1-one

OR305292-1G
APOSOR305292-1GEA 92.8 EUR
APOSOR305292-1G
6-Chloro-2,3-dihydro-1H-isoindol-1-one
6-Chloro-2,3-dihydro-1H-isoindol-1-one
Formel: C₈H₆ClNO
Molecular Weight: 167,592 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08234734
CAS-Nummer: 58083-59-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR