6-Brom-m-toluylsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR3053-1G
OR3053-5G
APOSOR3053-1GEA
21.9
EUR
APOSOR3053-1G
APOSOR3053-5G
6-Brom-m-toluylsäure 98%
6-Brom-m-toluylsäure
Formel:
C₈H₇BrO₂ Molecular Weight: 215.05 g/mol Schmelzpunkt: 136…140 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00079722 CAS-Nummer: 6967-82-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...