7-Methoxy-2,3-dihydro-1H-isoindol-1-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Methoxyisoindolin-1-one

OR305300-250MG
APOSOR305300-250MGEA 177 EUR
APOSOR305300-250MG
7-Methoxy-2,3-dihydro-1H-isoindol-1-one
7-Methoxy-2,3-dihydro-1H-isoindol-1-one
Formel: C₉H₉NO₂
Molecular Weight: 163,173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11215517
CAS-Nummer: 934389-18-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR