5-Methyl-1H-pyrazolo[4,3-b]pyridin-7-ol

Lieferant: Apollo Scientific

OR305331-250MG
APOSOR305331-250MGEA 165 EUR
APOSOR305331-250MG
5-Methyl-1H-pyrazolo[4,3-b]pyridin-7-ol
5-Methyl-1H-pyrazolo[4,3-b]pyridin-7-ol
Formel: C₇H₇N₃O
Molecular Weight: 149,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08236754
CAS-Nummer: 268547-52-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR