6-Bromo-1H-pyrazolo[4,3-c]pyridine

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Bromo-5-azaindazole

OR305335-250MG
APOSOR305335-250MGEA 55.1 EUR
APOSOR305335-250MG
6-Bromo-1H-pyrazolo[4,3-c]pyridine
6-Bromo-1H-pyrazolo[4,3-c]pyridine
Formel: C₆H₄BrN₃
Molecular Weight: 198,02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17010106
CAS-Nummer: 1206973-12-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR