Ethyl 2-nitrophenylacetate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2-(2-nitrophenyl)acetate, 2-Nitrophenylacetic acid ethyl ester

OR30534-1G
APOSOR30534-1GEA 30.6 EUR
APOSOR30534-1G
Ethyl 2-nitrophenylacetate 98%
Ethyl 2-nitrophenylacetate
Formel: C₁₀H₁₁NO₄
Molecular Weight: 209,199 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00092226
CAS-Nummer: 31912-02-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR