tert-Butyl (4-oxopiperidin-1-yl)acetate

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 2-(4-oxopiperidin-1-yl)acetate

OR305345-250MG
APOSOR305345-250MGEA 60.9 EUR
APOSOR305345-250MG
tert-Butyl (4-oxopiperidin-1-yl)acetate
tert-Butyl (4-oxopiperidin-1-yl)acetate
Formel: C₁₁H₁₉NO₃
Molecular Weight: 213,273 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07371646
CAS-Nummer: 149554-03-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR