1H-Pyrrolo[3,2-c]pyridine-4-carboxylic acid

Lieferant: Apollo Scientific

OR305346-250MG
APOSOR305346-250MGEA 331 EUR
APOSOR305346-250MG
1H-Pyrrolo[3,2-c]pyridine-4-carboxylic acid
1H-Pyrrolo[3,2-c]pyridine-4-carboxylic acid
Formel: C₈H₆N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09965904
CAS-Nummer: 1040682-84-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR