1H-Pyrrolo[2,3-b]pyridin-2-ol

Lieferant: Apollo Scientific
Achtung

OR305354-250MG
APOSOR305354-250MGEA 53.8 EUR
APOSOR305354-250MG
1H-Pyrrolo[2,3-b]pyridin-2-ol
1H-Pyrrolo[2,3-b]pyridin-2-ol
Formel: C₇H₆N₂O
Molecular Weight: 134,135 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18414674
CAS-Nummer: 1261802-66-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR