7-Bromo-1H-pyrazolo[3,4-c]pyridine
Lieferant: Apollo Scientific
|
Achtung
|
OR305363-1G
APOSOR305363-1GEA
1050
EUR
APOSOR305363-1G
7-Bromo-1H-pyrazolo[3,4-c]pyridine
7-Bromo-1H-pyrazolo[3,4-c]pyridine
Formel:
C₆H₄BrN₃ Molecular Weight: 198,02 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09265707 CAS-Nummer: 957760-11-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...